siRNA Details
Gene Symbol: Comp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016685.2 | NP_057894.2 | 10 | 1088 | ||
GenBank mRNA | AF033530.1 | - | 10 | 1078 | ||
BC128368.1 | - | 10 | 1078 | 40111643 | ||
BC128369.1 | - | 10 | 1077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
