siRNA Details
Gene Symbol: Copa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009938.4 | NP_034068.3 | 13 | 1550 | ||
GenBank mRNA | AK043479.1 | - | 13 | 1276 | ||
AK200292.1 | - | 2 | 93 | |||
BC024070.1 | - | 13 | 1203 | 5097228 | ||
BC047429.1 | - | 13 | 1241 | 3498656 | ||
BC082785.1 | - | 9 | 890 |
