siRNA Details
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053770.1 | - | 3 | 373 | ||
AF080580.1 | - | 4 | 436 | |||
AF098949.1 | - | 4 | 502 | |||
AK137841.1 | - | 2 | 2467 | |||
AK158380.1 | - | 4 | 487 | |||
BC028276.1 | - | 2 | 178 | |||
BC038681.1 | - | 4 | 469 | 1380996 | ||
U81277.1 | - | 3 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
