siRNA Details
Gene Symbol: Cox5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007747.2 | NP_031773.2 | 3, 4 | 419 | ||
GenBank mRNA | AK018723.1 | - | 3, 4 | 354 | ||
BC003831.1 | - | 2, 3 | 1545 | |||
BC034302.1 | - | 3, 4 | 372 | 5372849 | ||
X15963.1 | - | 3, 4 | 343 |

More Information
Other Species Targeted

Gene Ontology Categories:
