siRNA Details
Gene Symbol: Cox5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009942.2 | NP_034072.2 | 3 | 347 | ||
GenBank mRNA | AK008339.1 | - | 3 | 268 | ||
AK131673.1 | - | 3 | 268 | |||
AK135174.1 | - | 3 | 276 | |||
AK215114.1 | - | 1 | 52 | |||
BC005564.1 | - | 287 | ||||
BC096048.1 | - | 3 | 262 | 3465479 | ||
BC099474.1 | - | 3 | 276 | 30933585 | ||
CJ075429.1 | - | 3 | 352 | |||
X53157.1 | - | 3 | 256 |
Gene Symbol: Gm11273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023228.1 | - | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
