siRNA Details
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007749.3 | NP_031775.1 | 3 | 381 | ||
GenBank mRNA | AK002494.1 | - | 3 | 348 | ||
AK027989.1 | - | 3 | 343 | |||
AK131871.1 | - | 3 | 377 | |||
AK153444.1 | - | 3 | 353 | |||
AK168077.1 | - | 3 | 369 | |||
BB696274.1 | - | 3 | 392 | |||
BC010772.1 | - | 3 | 363 | 4216583 | ||
BC086792.1 | - | 3 | 353 | 6411273 | ||
X52940.1 | - | 3 | 347 |

More Information
Other Species Targeted

Gene Ontology Categories:
