siRNA Details
Gene Symbol: Cpox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007757.2 | NP_031783.2 | 6 | 1452 | ||
GenBank mRNA | AK166512.1 | - | 6 | 1315 | ||
BC017680.1 | - | 6 | 1309 | 4241687 | ||
D16333.1 | - | 6 | 1366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
