siRNA Details
Gene Symbol: Crabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013496.3 | NP_038524.1 | 3 | 426 | ||
GenBank mRNA | AK045283.1 | - | 3 | 368 | ||
AK144397.1 | - | 3 | 378 | |||
M31552.1 | - | 2 | 79 | |||
X15481.1 | - | 3 | 352 | |||
X15789.1 | - | 3 | 368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
