siRNA Details
Gene Symbol: Crabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284507.1 | NP_001271436.1 | 1 | 200 | ||
NM_013496.3 | NP_038524.1 | 1 | 200 | |||
GenBank mRNA | AK045283.1 | - | 1 | 142 | ||
AK144397.1 | - | 1 | 152 | |||
BC065787.1 | - | 1 | 128 | 891192 | ||
X15481.1 | - | 1 | 126 | |||
X15789.1 | - | 1 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
