siRNA Details
Gene Symbol: Crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007761.2 | NP_031787.1 | 3, 4 | 247 | ||
GenBank mRNA | AF028242.1 | - | 3, 4 | 237 | ||
AF118271.1 | - | 3, 4 | 247 | |||
AK075587.1 | - | 3, 4 | 242 | |||
BC012676.1 | - | 3, 4 | 181 | 3989860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
