siRNA Details
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009952.2 | NP_034082.1 | 2 | 271 | ||
NM_133828.2 | NP_598589.2 | 2 | 271 | |||
GenBank mRNA | AF448507.1 | - | 2 | 147 | ||
AF448508.1 | - | 799 | ||||
AK042595.1 | - | 2 | 244 | |||
AK044992.1 | - | 2 | 86 | |||
AK044993.1 | - | 2 | 86 | |||
AK079223.1 | - | 2 | 267 | |||
BC021649.1 | - | 2 | 204 | 4017170 | ||
BY782293.1 | - | 2 | 228 | |||
M95106.1 | - | 2 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
