siRNA Details
Gene Symbol: Crem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110850.2 | NP_001104320.1 | 3 | 229 | ||
NM_001110851.1 | NP_001104321.1 | 4 | 488 | |||
NM_001110852.1 | NP_001104322.1 | 3 | 341 | |||
NM_001110853.2 | NP_001104323.1 | 3 | 267 | |||
NM_001110856.2 | NP_001104326.1 | 4 | 376 | |||
NM_001110857.2 | NP_001104327.1 | 3 | 229 | |||
NM_001110858.2 | NP_001104328.1 | 3 | 229 | |||
NM_001110859.2 | NP_001104329.1 | 4 | 376 | |||
NM_001271503.1 | NP_001258432.1 | 4 | 376 | |||
NM_001271504.1 | NP_001258433.1 | 4 | 376 | |||
NM_001271505.1 | NP_001258434.1 | 3 | 229 | |||
NM_001271506.1 | NP_001258435.1 | 4 | 376 | |||
NM_013498.3 | NP_038526.2 | 3 | 229 | |||
GenBank mRNA | AI461618.1 | - | 4 | 338 | ||
AK009841.1 | - | 3 | 261 | |||
AK010806.1 | - | 3 | 342 | |||
AK015641.1 | - | 3 | 268 | |||
AK016156.1 | - | 3 | 254 | |||
AK076797.1 | - | 3 | 190 | |||
AK161366.1 | - | 4 | 422 | |||
AY738715.1 | - | 3 | 292 | |||
AY738716.1 | - | 2 | 145 | |||
AY738717.1 | - | 3 | 292 | |||
AY738718.1 | - | 3 | 292 | |||
AY738719.1 | - | 2 | 145 | |||
AY738720.1 | - | 4 | 388 | |||
AY738721.1 | - | 3 | 292 | |||
BC034856.1 | - | 2 | 138 | |||
M60285.1 | - | 3 | 157 | |||
S64590.1 | - | 3 | 274 |

More Information
Other Species Targeted

Gene Ontology Categories:
