siRNA Details
Gene Symbol: Crh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205769.2 | NP_991338.1 | 2 | 420 | ||
GenBank mRNA | BC116921.1 | - | 1 | 202 | 40126240 | |
BC119036.1 | - | 1 | 202 | 8733785 |

More Information
Other Species Targeted

Gene Ontology Categories:
