siRNA Details
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198839.1 | NP_001185768.1 | 2 | 556 | ||
NM_009954.3 | NP_034084.2 | 2 | 619 | |||
GenBank mRNA | AK138372.1 | - | 2 | 526 | ||
AK145863.1 | - | 2 | 620 | |||
AK159168.1 | - | 2 | 523 | |||
AK159297.1 | - | 2 | 523 | |||
AK166248.1 | - | 2 | 494 | |||
AK167352.1 | - | 2 | 557 | |||
AK167504.1 | - | 2 | 557 | |||
AK167516.1 | - | 2 | 620 | |||
AK167717.1 | - | 2 | 620 | |||
AK213855.1 | - | 1 | 184 | |||
BC057578.1 | - | 2 | 579 | 6401186 | ||
U28151.1 | - | 2 | 530 | |||
U48853.1 | - | 2 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
