siRNA Details
Gene Symbol: Crx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113330.1 | NP_001106801.1 | 4 | 974 | ||
NM_007770.4 | NP_031796.1 | 4 | 1002 | |||
GenBank mRNA | AK053533.1 | - | 4 | 944 | ||
AK139691.1 | - | 4 | 977 | |||
BC016502.1 | - | 4 | 916 | 4527863 | ||
U77615.1 | - | 4 | 955 |
