siRNA Details
Gene Symbol: Cryba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009965.2 | NP_034095.1 | 2, 3 | 115 | ||
GenBank mRNA | AJ239052.1 | - | 2, 3 | 115 | ||
BC107342.2 | - | 2, 3 | 115 | 40055280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
