siRNA Details
Gene Symbol: Cryba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021541.3 | NP_067516.1 | 4 | 445 | ||
GenBank mRNA | AJ272227.1 | - | 4 | 437 | ||
AK021374.1 | - | 4 | 419 | |||
AY160973.1 | - | 4 | 410 | |||
BC052746.1 | - | 4 | 408 | 6824801 | ||
BC056989.1 | - | 4 | 399 | 6824245 | ||
CF740624.1 | - | 4 | 444 |

More Information
Other Species Targeted

Gene Ontology Categories:
