siRNA Details
Gene Symbol: Crygf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027010.2 | NP_081286.1 | 3 | 323 | ||
GenBank mRNA | AK013953.1 | - | 3 | 324 | ||
BC056453.1 | - | 3 | 314 | 6824171 | ||
M64543.1 | - | 2 | 291 |
Gene Symbol: Cryge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007777.3 | NP_031803.3 | 3 | 337 | ||
GenBank mRNA | AK014301.1 | - | 3 | 324 | ||
BC057012.1 | - | 312 | ||||
BC078417.1 | - | 312 | 6824205 | |||
BC132506.1 | - | 3 | 337 | 40130783 | ||
BC132508.1 | - | 3 | 337 | 40130785 |

More Information
Other Species Targeted

Gene Ontology Categories:
