siRNA Details
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113530.1 | NP_001107002.1 | 6 | 1083 | ||
NM_007778.4 | NP_031804.3 | 6 | 1083 | |||
GenBank mRNA | AK136808.1 | - | 6 | 857 | ||
AK138489.1 | - | 6 | 857 | |||
AK154261.1 | - | 6 | 1084 | |||
AK154872.1 | - | 6 | 857 | |||
AK160995.1 | - | 6 | 857 | |||
AK166370.1 | - | 6 | 856 | |||
AK170154.1 | - | 6 | 793 | |||
BC066187.1 | - | 6 | 837 | 30106218 | ||
BC066200.1 | - | 6 | 837 | 30100524 | ||
BC066205.1 | - | 6 | 837 | 30100601 | ||
M21149.1 | - | 6 | 746 | |||
M21952.1 | - | 6 | 836 | |||
X05010.1 | - | 6 | 858 |

More Information
Other Species Targeted

Gene Ontology Categories:
