siRNA Details
Gene Symbol: Csf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009969.4 | NP_034099.2 | 4 | 660 | ||
GenBank mRNA | AK053196.1 | - | 4 | 660 | ||
BC116878.1 | - | 4 | 394 | 40126197 | ||
BC116880.1 | - | 4 | 394 | 40126199 | ||
EU366957.1 | - | 4 | 371 | |||
M11848.1 | - | 4 | 329 | |||
X02333.1 | - | 4 | 544 | |||
X03019.1 | - | 4 | 403 | |||
X03221.1 | - | 4 | 402 | |||
X05906.1 | - | 4 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
