siRNA Details
Gene Symbol: Csn1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286015.1 | NP_001272944.1 | 31 | 917 | ||
NM_001286016.1 | NP_001272945.1 | 32 | 884 | |||
NM_007784.3 | NP_031810.1 | 32 | 941 | |||
GenBank mRNA | AK052787.1 | - | 1 | 373 | ||
AK085690.1 | - | 32 | 945 | |||
BC002101.1 | - | 32 | 853 | 3484624 | ||
BC003859.1 | - | 32 | 925 | 3486384 | ||
BC006024.1 | - | 31 | 901 | 3592625 | ||
BC007192.1 | - | 32 | 925 | |||
BC008278.1 | - | 32 | 868 | 2812523 | ||
BC040246.1 | - | 32 | 880 | 2647887 | ||
M36780.1 | - | 31 | 880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
