siRNA Details
Gene Symbol: Csn1s2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142688.1 | - | 11, 12 | 478 | ||
AK164738.1 | - | 11, 12 | 482 | |||
BC139326.1 | - | 11, 12 | 458 | 8862348 | ||
D10215.1 | - | 11, 12 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
