siRNA Details
Gene Symbol: Dnajc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271584.1 | NP_001258513.1 | 3 | 335 | ||
NM_001271585.1 | NP_001258514.1 | 3 | 362 | |||
NM_016775.3 | NP_058055.1 | 2 | 282 | |||
GenBank mRNA | AF032115.1 | - | 1 | 58 | ||
AK007879.1 | - | 2 | 225 | |||
AK029006.1 | - | 2 | 220 | |||
AK032373.1 | - | 2 | 214 | |||
AK083584.1 | - | 2 | 243 | |||
BC012268.1 | - | 3 | 252 | 3500124 | ||
BC018231.1 | - | 2 | 209 | |||
BY101584.1 | - | 3 | 270 | |||
BY333630.1 | - | 2 | 282 | |||
CB249513.1 | - | 3 | 249 | |||
CN459638.1 | - | 2 | 98 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
