siRNA Details
Gene Symbol: Csrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007792.4 | NP_031818.3 | 4 | 404 | ||
GenBank mRNA | AF037208.1 | - | 4 | 404 | ||
AK003328.1 | - | 4 | 403 | |||
AK010626.1 | - | 4 | 405 | |||
AK012647.1 | - | 4 | 402 | |||
BC012663.1 | - | 4 | 389 | 4210909 | ||
CK031156.1 | - | 4 | 390 | |||
D88792.1 | - | 3 | 330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
