siRNA Details
Gene Symbol: Cst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009976.3 | NP_034106.2 | 3 | 571 | ||
GenBank mRNA | AK002438.1 | - | 3 | 572 | ||
AK013676.1 | - | 3 | 540 | |||
AK014368.1 | - | 3 | 591 | |||
AK131728.1 | - | 3 | 589 | |||
AK146333.1 | - | 3 | 600 | |||
AK151160.1 | - | 3 | 536 | |||
AK161856.1 | - | 3 | 534 | |||
AK166430.1 | - | 3 | 563 | |||
AK212642.1 | - | 3 | 288 | |||
BC002072.1 | - | 3 | 517 | 3593373 | ||
M59470.1 | - | 3 | 504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
