siRNA Details
Gene Symbol: Cstb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007793.3 | NP_031819.1 | 2 | 212 | ||
GenBank mRNA | AK002403.1 | - | 2 | 176 | ||
AK151205.1 | - | 2 | 142 | |||
BC056170.1 | - | 2 | 165 | 5713930 | ||
BY091749.1 | - | 2 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
