siRNA Details
Gene Symbol: Ctla2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145799.1 | NP_001139271.1 | 2 | 184 | ||
NM_007796.2 | NP_031822.2 | 2 | 222 | |||
GenBank mRNA | AA451288.1 | - | 2 | 184 | ||
AK005471.1 | - | 2 | 159 | |||
AK018801.1 | - | 2 | 177 | |||
AK132290.1 | - | 2 | 155 | |||
AK167433.1 | - | 2 | 155 | |||
AK167664.1 | - | 2 | 156 | |||
AY034577.1 | - | 2 | 131 | |||
BC028437.1 | - | 2 | 148 | 1244311 | ||
X15591.1 | - | 2 | 131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
