siRNA Details
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163159.1 | NP_001156631.1 | 4 | 589 | ||
NM_001163160.1 | NP_001156632.1 | 4 | 523 | |||
NM_009981.4 | NP_034111.1 | 4 | 425 | |||
GenBank mRNA | AK044585.1 | - | 4 | 426 | ||
AK076050.1 | - | 4 | 350 | |||
AK076830.1 | - | 4 | 524 | |||
BB642590.1 | - | 4 | 426 | |||
BC018313.1 | - | 3 | 308 | 4162053 | ||
L28956.1 | - | 4 | 394 | |||
Z12302.1 | - | 4 | 384 |

More Information
Other Species Targeted

Gene Ontology Categories:
