siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 3 | 415 | ||
NM_001244903.1 | NP_001231832.1 | 2 | 362 | |||
NM_017368.3 | NP_059064.2 | 3 | 514 | |||
NM_198683.2 | NP_941955.1 | 2 | 362 | |||
GenBank mRNA | AF267535.1 | - | 1 | 118 | ||
AJ007987.1 | - | 1 | 107 | |||
AK014492.1 | - | 2 | 275 | |||
AK143698.1 | - | 3 | 316 | |||
AK156725.1 | - | 2 | 275 | |||
AK161213.1 | - | 2 | 322 | |||
AK214229.1 | - | 1 | 86 | |||
BC021393.1 | - | 2 | 227 | |||
BU056861.1 | - | 2 | 362 | |||
BU923429.1 | - | 2 | 217 | |||
CJ092744.1 | - | 3 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
