siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 6 | 715 | ||
NM_001244903.1 | NP_001231832.1 | 5 | 662 | |||
NM_017368.3 | NP_059064.2 | 6 | 814 | |||
NM_198683.2 | NP_941955.1 | 5 | 662 | |||
GenBank mRNA | AB050499.1 | - | 2 | 123 | ||
AF267535.1 | - | 4 | 418 | |||
AF314172.1 | - | 4 | 256 | |||
AJ007987.1 | - | 4 | 407 | |||
AK014492.1 | - | 5 | 575 | |||
AK143698.1 | - | 6 | 616 | |||
AK156725.1 | - | 5 | 575 | |||
AK161213.1 | - | 5 | 622 | |||
BC021393.1 | - | 5 | 527 | |||
BU923429.1 | - | 5 | 517 | |||
X61451.1 | - | 1 | 110 |

More Information
Other Species Targeted

Gene Ontology Categories:
