siRNA Details
Gene Symbol: Cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025192.3 | NP_001020363.1 | 3 | 437 | ||
NM_001276263.1 | NP_001263192.1 | 3 | 437 | |||
NM_009988.4 | NP_034118.1 | 3 | 437 | |||
GenBank mRNA | AK004908.2 | - | 3 | 362 | ||
AK145569.1 | - | 3 | 363 | |||
AK160457.1 | - | 3 | 359 | |||
BC004680.1 | - | 3 | 349 | |||
BC016457.1 | - | 3 | 352 | 4216727 | ||
BQ895050.1 | - | 434 | ||||
U90715.1 | - | 3 | 337 | |||
Y10320.1 | - | 3 | 276 | |||
Y11929.1 | - | 3 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
