siRNA Details
Gene Symbol: Cyb561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007805.4 | NP_031831.2 | 4 | 436 | ||
GenBank mRNA | AK014395.2 | - | 4 | 437 | ||
AK028780.1 | - | 3 | 310 | |||
AK158978.1 | - | 5 | 767 | |||
AK169711.1 | - | 4 | 425 | |||
BC006732.1 | - | 4 | 476 | 3963612 | ||
U16297.1 | - | 4 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
