siRNA Details
Gene Symbol: Cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007809.3 | NP_031835.3 | 3 | 655 | ||
GenBank mRNA | AK132830.1 | - | 3 | 549 | ||
AK134376.1 | - | 3 | 549 | |||
BC064793.1 | - | 3 | 629 | 30436278 | ||
M64863.1 | - | 3 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
