siRNA Details
Gene Symbol: Cyp19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007810.3 | NP_031836.1 | 6 | 796 | ||
GenBank mRNA | AK143323.1 | - | 6 | 876 | ||
BC103670.1 | - | 6 | 779 | 40047093 | ||
BC104489.1 | - | 6 | 779 | 40047095 | ||
D00659.1 | - | 6 | 801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
