siRNA Details
Gene Symbol: Cyp19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007810.3 | NP_031836.1 | 6 | 873 | ||
GenBank mRNA | AK143323.1 | - | 6 | 953 | ||
BC103670.1 | - | 6 | 856 | 40047093 | ||
BC104489.1 | - | 6 | 856 | 40047095 | ||
D00659.1 | - | 6 | 878 |

More Information
Other Species Targeted

Gene Ontology Categories:
