siRNA Details
Gene Symbol: Cyp26a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007811.2 | NP_031837.2 | 7 | 1323 | ||
GenBank mRNA | AA239785.1 | - | 3 | 252 | ||
AF115769.1 | - | 7 | 1247 | |||
BC012673.1 | - | 7 | 1283 | 4210893 | ||
Y12657.1 | - | 7 | 1277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
