siRNA Details
Gene Symbol: Cyp2c40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010004.2 | NP_034134.2 | 6 | 1001 | ||
GenBank mRNA | AF047727.1 | - | 6 | 915 | ||
BC139471.1 | - | 6 | 1001 | 8862493 | ||
BC139472.1 | - | 6 | 1001 | 8862494 |
Gene Symbol: Cyp2c68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039555.2 | NP_001034644.2 | 6 | 969 | ||
GenBank mRNA | AK136413.1 | - | 5 | 599 |
Gene Symbol: Cyp2c69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104525.1 | NP_001097995.1 | 6 | 971 | ||
GenBank mRNA | BC145630.1 | - | 7 | 992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
