siRNA Details
Gene Symbol: Cyp2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021282.2 | NP_067257.1 | 1 | 166 | ||
GenBank mRNA | BC013451.1 | - | 1 | 166 | 4223797 | |
BC042693.1 | - | 1 | 145 | |||
DQ838976.1 | - | 1 | 153 | |||
L11650.1 | - | 1 | 259 | |||
X62595.1 | - | 1 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
