siRNA Details
Gene Symbol: Cyp46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010010.1 | NP_034140.1 | 2, 3 | 268 | ||
GenBank mRNA | AF094479.1 | - | 2, 3 | 268 | ||
AK140740.1 | - | 2, 3 | 273 | |||
BC018307.1 | - | 2, 3 | 291 | 4195579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
