siRNA Details
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008702.2 | NP_001008702.1 | 11, 12 | 1681 | ||
NM_001037905.3 | NP_001032994.1 | 11, 12 | 1755 | |||
NM_001102400.1 | NP_001095870.1 | 10, 11 | 1692 | |||
NM_023118.5 | NP_075607.2 | 12, 13 | 2409 | |||
GenBank mRNA | AK162744.1 | - | 10, 11 | 1663 | ||
AK162851.1 | - | 11, 12 | 1726 | |||
AK165519.1 | - | 11, 12 | 1639 | |||
AK165810.1 | - | 11, 12 | 1636 | |||
AK168375.1 | - | 11, 12 | 1728 | |||
BC006588.1 | - | 10, 11 | 1576 | 3587226 | ||
BC016887.1 | - | 11, 12 | 1621 | 4242174 | ||
U18869.1 | - | 12, 13 | 2342 |

More Information
Other Species Targeted

Gene Ontology Categories:
