siRNA Details
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008702.2 | NP_001008702.1 | 6 | 686 | ||
NM_001037905.3 | NP_001032994.1 | 6 | 760 | |||
NM_001102400.1 | NP_001095870.1 | 6 | 760 | |||
NM_023118.5 | NP_075607.2 | 6 | 760 | |||
GenBank mRNA | AK002850.1 | - | 6 | 643 | ||
AK154716.1 | - | 6 | 744 | |||
AK162744.1 | - | 6 | 731 | |||
AK162851.1 | - | 6 | 731 | |||
AK165519.1 | - | 6 | 644 | |||
AK165810.1 | - | 6 | 641 | |||
AK168375.1 | - | 6 | 734 | |||
BC006588.1 | - | 5 | 581 | 3587226 | ||
BC016887.1 | - | 6 | 626 | 4242174 | ||
CD348957.1 | - | 4 | 283 |

More Information
Other Species Targeted

Gene Ontology Categories:
