siRNA Details
Gene Symbol: Dad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010015.4 | NP_034145.1 | 2 | 870 | ||
GenBank mRNA | AK030925.1 | - | 2 | 820 | ||
AK054480.1 | - | 2 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
