siRNA Details
Gene Symbol: Daf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007827.2 | NP_031853.2 | 10 | 1147 | ||
GenBank mRNA | BC115954.1 | - | 10 | 1147 | ||
BC127150.1 | - | 8 | 818 | 40055987 | ||
L41365.1 | - | 10 | 1156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
