siRNA Details
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016811.2 | NP_058091.2 | 23, 24 | 2449 | ||
GenBank mRNA | AF085219.1 | - | 22, 23 | 2135 | ||
AK008063.1 | - | 11, 12 | 1108 | |||
AK008272.1 | - | 12, 13 | 1116 | |||
AK008314.1 | - | 12, 13 | 1116 | |||
BC006713.1 | - | 23, 24 | 2449 | 3584592 | ||
JN832677.1 | - | 23, 24 | 2392 | |||
JN832678.1 | - | 23, 24 | 2744 | |||
JN832679.1 | - | 23, 24 | 2468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
