siRNA Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010019.3 | NP_034149.2 | 5 | 593 | ||
GenBank mRNA | AB018002.1 | - | 5 | 583 | ||
AF052942.1 | - | 3 | 273 | |||
AK004350.1 | - | 5 | 566 | |||
BC022165.1 | - | 5 | 562 | 4016911 | ||
CK032192.1 | - | 5 | 626 | |||
GU109477.1 | - | 5 | 593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
