siRNA Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010019.3 | NP_034149.2 | 5 | 590 | ||
GenBank mRNA | AB018002.1 | - | 5 | 580 | ||
AF052942.1 | - | 3 | 270 | |||
AK004350.1 | - | 5 | 563 | |||
BC022165.1 | - | 5 | 559 | 4016911 | ||
CK032192.1 | - | 5 | 623 | |||
GU109477.1 | - | 5 | 590 |
