siRNA Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010019.3 | NP_034149.2 | 5 | 664 | ||
GenBank mRNA | AB018002.1 | - | 5 | 654 | ||
AF052942.1 | - | 3 | 344 | |||
AK004350.1 | - | 5 | 637 | |||
BC022165.1 | - | 5 | 633 | 4016911 | ||
BX527735.1 | - | 1 | 48 | |||
GU109477.1 | - | 5 | 664 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013109, XM_006243308, XM_008766241, XM_006243306, XM_006243309, and others.
Gene Ontology Categories:
