siRNA Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199733.1 | NP_001186662.1 | 6 | 2200 | ||
NM_007829.4 | NP_031855.3 | 6 | 2100 | |||
GenBank mRNA | AF006040.1 | - | 5 | 1951 | ||
AK090356.1 | - | 7 | 1911 | |||
AK133863.1 | - | 6 | 2008 | |||
AK144332.1 | - | 6 | 2033 | |||
AK145906.1 | - | 6 | 2031 | |||
AK146743.1 | - | 6 | 2027 | |||
AK151279.1 | - | 6 | 2021 | |||
AK153149.1 | - | 6 | 1988 | |||
AK156425.1 | - | 6 | 2198 | |||
AK172616.1 | - | 6 | 2201 | |||
AY734459.1 | - | 4 | 1664 | |||
AY734464.1 | - | 1 | 273 | |||
AY734465.1 | - | 4 | 1575 | |||
AY734466.1 | - | 1855 | ||||
AY758274.1 | - | 1 | 406 | |||
BC128373.1 | - | 6 | 2010 | 40111681 | ||
KJ650207.1 | - | 5 | 1927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
