siRNA Details
Gene Symbol: Dbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016974.3 | NP_058670.2 | 4 | 1202 | ||
GenBank mRNA | AK140243.1 | - | 4 | 1079 | ||
AK178288.1 | - | 2 | 199 | |||
AK184638.1 | - | 1 | 46 | |||
AK213757.1 | - | 1 | 45 | |||
BC018323.1 | - | 4 | 1165 | 4195116 | ||
BC064094.1 | - | 2 | 1431 | |||
BC084676.1 | - | 4 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
