siRNA Details
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111051.1 | NP_001104521.1 | 10 | 1218 | ||
NM_001111052.1 | NP_001104522.1 | 10 | 1218 | |||
NM_001195538.1 | NP_001182467.1 | 14 | 2079 | |||
NM_001195539.1 | NP_001182468.1 | 10 | 1266 | |||
NM_019978.3 | NP_064362.1 | 14 | 2127 | |||
GenBank mRNA | AB093234.1 | - | 14 | 2033 | ||
AF155819.1 | - | 14 | 2025 | |||
AF155821.1 | - | 10 | 1077 | |||
AK032424.1 | - | 10 | 1108 | |||
AK047716.1 | - | 10 | 1005 | |||
AK081200.1 | - | 10 | 1061 | |||
AY968047.1 | - | 10 | 966 | |||
AY968048.1 | - | 10 | 969 | |||
BC133685.1 | - | 14 | 1881 | 40110683 | ||
BQ930085.1 | - | 2 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
